Assay Details
Gene Symbol: Ttc39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027238.2 | NP_081514.1 | 19 - 20 | 1836 | 140 | |
GenBank mRNA | AK037208.1 | - | 19 - 20 | 1837 | 140 | |
AK043383.1 | - | 2 - 3 | 574 | 140 | ||
AK132655.1 | - | 14 - 15 | 1287 | 140 | ||
BC151021.1 | - | 19 - 20 | 1798 | 9087934 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
