Assay Details
Gene Symbol: Ttc39b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027238.2 | NP_081514.1 | 3 - 4 | 262 | 61 | |
GenBank mRNA | AK007856.1 | - | 3 - 4 | 263 | 61 | |
AK037208.1 | - | 3 - 4 | 263 | 61 | ||
AK049108.1 | - | 3 - 4 | 313 | 61 | ||
BC151021.1 | - | 3 - 4 | 224 | 9087934 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
