Assay Details
Gene Symbol: Stam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019667.2 | NP_062641.1 | 7 - 8 | 755 | 91 | |
GenBank mRNA | AB012611.1 | - | 7 - 8 | 770 | 91 | |
AK004604.2 | - | 7 - 8 | 755 | 91 | ||
AK035789.1 | - | 808 | 91 | |||
AK044230.1 | - | 6 - 7 | 684 | 91 | ||
AK078244.1 | - | 7 - 8 | 787 | 91 | ||
AK159699.1 | - | 7 - 8 | 784 | 91 | ||
AK168729.1 | - | 7 - 8 | 787 | 91 | ||
BC013818.1 | - | 7 - 8 | 791 | 3671426 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
