Assay Details
Gene Symbol: Sec24c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168273.1 | NP_001161745.1 | 18 - 19 | 2299 | 63 | |
NM_172596.2 | NP_766184.2 | 17 - 18 | 2527 | 63 | ||
GenBank mRNA | AK036519.1 | - | 17 - 18 | 2527 | 63 | |
AK122199.1 | - | 18 - 19 | 2277 | 63 | ||
AK132090.1 | - | 12 - 13 | 1554 | 63 | ||
BC027157.1 | - | 4 - 5 | 453 | 63 | ||
BC027775.1 | - | 8 - 9 | 792 | 63 | ||
BC040370.1 | - | 17 - 18 | 2505 | 4239204 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
