Assay Details
Gene Symbol: Itgax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021334.2 | NP_067309.1 | 26 - 27 | 3137 | 65 | |
GenBank mRNA | AF211864.1 | - | 26 - 27 | 3137 | 65 | |
AK155816.1 | - | 26 - 27 | 3159 | 65 | ||
AK170432.1 | - | 26 - 27 | 3155 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
