Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006547.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006547.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006547.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006563.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006563.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006563.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006541.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006541.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006541.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006552.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006552.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006552.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006555.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006555.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006555.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006561.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006561.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006561.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006559.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006559.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006559.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006558.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006558.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006558.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006562.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006562.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006562.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006556.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006556.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006556.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006560.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006560.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006560.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006551.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006551.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006551.mm.1 |
NM_029924.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006553.mm.1 |
TC0200000783.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006553.mm.1 |
NM_029924.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006553.mm.1 |
NM_029924.2 |
Refseq |