Assay Details
Gene Symbol: Mbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290656.1 | NP_001277585.1 | 4 - 5 | 2721 | 78 | |
NM_029924.2 | NP_084200.1 | 11 - 12 | 3785 | 78 | ||
GenBank mRNA | AK173176.1 | - | 4 - 5 | 2652 | 78 | |
BC050251.1 | - | 1 - 2 | 1746 | 78 | ||
BC138375.1 | - | 4 - 5 | 2626 | 8861395 | 78 | |
BC138378.1 | - | 4 - 5 | 2626 | 8861398 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
