Assay Details
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013469.2 | NP_038497.2 | 10 - 11 | 1228 | 71 | |
GenBank mRNA | AK044203.1 | - | 10 - 11 | 1201 | 71 | |
AK086039.1 | - | 2 - 3 | 130 | 71 | ||
AK137158.1 | - | 9 - 10 | 1538 | 71 | ||
AK140375.1 | - | 10 - 11 | 1201 | 71 | ||
AK170850.1 | - | 8 - 9 | 2612 | 71 | ||
BC012875.1 | - | 10 - 11 | 1148 | 3500318 | 71 | |
CT010331.1 | - | 9 - 10 | 1079 | 71 | ||
U65986.1 | - | 10 - 11 | 1213 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
