Assay Details
Gene Symbol: Klc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029091.2 | NP_083367.1 | 8 - 9 | 1225 | 65 | |
GenBank mRNA | AK004774.1 | - | 8 - 9 | 1226 | 65 | |
AK170793.1 | - | 8 - 9 | 1209 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
