Assay Details
Gene Symbol: Slc12a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079690.1 | NP_001073158.1 | 7 - 8 | 1190 | 90 | |
NM_183354.2 | NP_899197.2 | 7 - 8 | 1190 | 90 | ||
GenBank mRNA | BC016888.1 | - | 6 - 7 | 1168 | 4236020 | 90 |
U20973.1 | - | 7 - 8 | 1188 | 90 | ||
U20974.1 | - | 7 - 8 | 1188 | 90 | ||
U20975.1 | - | 7 - 8 | 1188 | 90 | ||
U61381.1 | - | 6 - 7 | 1159 | 90 | ||
U94518.1 | - | 6 - 7 | 1137 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
