Assay Details
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286217.1 | NP_001273146.1 | 3 - 4 | 275 | 75 | |
NM_026928.3 | NP_081204.2 | 3 - 4 | 275 | 75 | ||
NR_104415.1 | 3 - 4 | 275 | 75 | |||
GenBank mRNA | AK007494.1 | - | 3 - 4 | 238 | 75 | |
AK042452.1 | - | 238 | 75 | |||
AK044731.1 | - | 3 - 4 | 240 | 75 | ||
AK079410.1 | - | 3 - 4 | 241 | 75 | ||
AK163708.1 | - | 3 - 4 | 179 | 75 | ||
BC028662.1 | - | 3 - 4 | 236 | 1332340 | 75 | |
CK618091.1 | - | 3 - 4 | 210 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
