Assay Details
Gene Symbol: Nkx2-2os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_030769.2 | 5 - 6 | 3989 | 67 | ||
GenBank mRNA | AK045921.1 | - | 5 - 6 | 3990 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817796 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119146 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818573 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 864108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 430098 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44716 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61699 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 233477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4110 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227328 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499305 | AK017451.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499305 | BC023133.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499305 | NM_027799.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 499305 | NM_146024.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278729 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908123 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940096 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580220 | AK033654.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580220 | AK049338.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580220 | AK153792.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580220 | BC062133.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 580220 | NM_172490.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 434561 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 524345 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552266 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 162288 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133439 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912679 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 105356 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230146 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621008 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740382 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 656778 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 418582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680114 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735663 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 764519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19050 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 231485 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18540 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858071 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 555594 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734437 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28550 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 878185 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575108 | AK053165.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575108 | BC059880.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575108 | NM_001098836.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575108 | NM_001098837.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 152964 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10484 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172000 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117452 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 109211 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125799 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104487 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149326 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730870 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00521314 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104487 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149326 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730794 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730764 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00521231 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04731014 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730895 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730700 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730750 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04730929 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00521419 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135135 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04730895 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00521231 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00521314 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104487 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149326 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P00521419 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01159024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02093194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723602 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02266158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01763013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723621 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723625 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01455838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279474 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01261336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02093200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02093188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02266155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01629817 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02200251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01817591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722887 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01385846 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723249 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02005137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02279237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722868 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723043 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01220696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01217392 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02200237 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723626 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723593 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723211 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01899458 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01278025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722881 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01492781 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01440141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01275802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01822188 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02100044 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01692863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723592 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723020 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01692865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01278016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723584 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723577 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723583 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01492782 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723258 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723263 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01944025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01180525 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01817600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02200245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723383 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723610 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01739093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01870957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801757 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01692864 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01343600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01449534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723210 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02093199 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723540 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01489710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723369 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723045 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723247 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01763011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723011 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723016 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723534 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723585 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723600 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723628 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02160205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01817599 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722923 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01899471 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01227888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21722863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723623 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723212 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01817601 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21723618 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01801755 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02200242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01763018 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0200017313.mm.1 | TC0200002180.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017313.mm.1 | NR_030769.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017315.mm.1 | TC0200002180.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017315.mm.1 | NR_030769.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200017314.mm.1 | TC0200002180.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200017314.mm.1 | NR_030769.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 875419 | - | - |
