Assay Details
Gene Symbol: Bag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013863.5 | NP_038891.4 | 2 - 3 | 797 | 80 | |
GenBank mRNA | AB041583.1 | - | 2 - 3 | 764 | 80 | |
AF130471.1 | - | 2 - 3 | 530 | 80 | ||
AK007414.1 | - | 2 - 3 | 608 | 80 | ||
AK016510.1 | - | 2 - 3 | 604 | 80 | ||
AK036675.1 | - | 2 - 3 | 606 | 80 | ||
AK171734.1 | - | 2 - 3 | 739 | 80 | ||
AK214932.1 | - | 1 - 2 | 347 | 80 | ||
BC145765.1 | - | 2 - 3 | 710 | 40131068 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
