Assay Details
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089086.1 | - | 21 - 22 | 2317 | 68 | |
AK134118.1 | - | 10 - 11 | 2364 | 68 | ||
AK150411.1 | - | 21 - 22 | 2319 | 68 | ||
AK151535.1 | - | 13 - 14 | 1548 | 68 | ||
AK152675.1 | - | 21 - 22 | 2336 | 68 | ||
AK152769.1 | - | 21 - 22 | 2318 | 68 | ||
AK153065.1 | - | 21 - 22 | 2318 | 68 | ||
AK155772.1 | - | 21 - 22 | 2317 | 68 | ||
AK155912.1 | - | 21 - 22 | 2337 | 68 | ||
AK160041.1 | - | 21 - 22 | 2320 | 68 | ||
AK171022.1 | - | 19 - 20 | 2894 | 68 | ||
AK171060.1 | - | 21 - 22 | 2317 | 68 | ||
BC025584.1 | - | 21 - 22 | 2371 | 5324943 | 68 | |
X13335.1 | - | 21 - 22 | 2316 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
