Assay Details
Gene Symbol: Ptk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175168.4 | NP_780377.1 | 11 - 12 | 1928 | 65 | |
GenBank mRNA | AF531873.1 | - | 11 - 12 | 1778 | 65 | |
AK053044.1 | - | 11 - 12 | 1861 | 65 | ||
AK081793.1 | - | 1 - 2 | 2075 | 65 | ||
AK163135.1 | - | 11 - 12 | 1861 | 65 | ||
BC076578.1 | - | 11 - 12 | 1928 | 30619280 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
