Assay Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146043.2 | NP_001139515.1 | 19 - 20 | 2135 | 62 | |
NM_001271463.1 | NP_001258392.1 | 19 - 20 | 2135 | 62 | ||
NM_001271464.1 | NP_001258393.1 | 18 - 19 | 2054 | 62 | ||
NM_001271465.1 | NP_001258394.1 | 19 - 20 | 2162 | 62 | ||
NM_177266.5 | NP_796240.3 | 19 - 20 | 2168 | 62 | ||
GenBank mRNA | AK035819.1 | - | 19 - 20 | 2073 | 62 | |
AK044371.1 | - | 18 - 19 | 2055 | 62 | ||
AK052633.1 | - | 3 - 4 | 478 | 62 | ||
AK083441.1 | - | 18 - 19 | 2174 | 62 | ||
AK161797.1 | - | 19 - 20 | 2071 | 62 | ||
BC057062.1 | - | 8 - 9 | 1946 | 62 | ||
BC138233.1 | - | 19 - 20 | 2066 | 8861252 | 62 | |
BC145208.1 | - | 19 - 20 | 2066 | 9053759 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
