Assay Details
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013024.2 | NP_001013042.1 | 15 - 16 | 2005 | 112 | |
GenBank mRNA | AK135141.1 | - | 15 - 16 | 2057 | 112 | |
BC090999.1 | - | 15 - 16 | 2005 | 6754549 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
