Assay Details
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AI790078.1 | - | 1 - 2 | 177 | 66 | |
AK154170.1 | - | 8 - 9 | 1861 | 66 | ||
BC029153.1 | - | 2 - 3 | 186 | 66 | ||
BC030904.1 | - | 7 - 8 | 1156 | 66 | ||
BC034078.1 | - | 10 - 11 | 1548 | 66 | ||
BC076622.1 | - | 7 - 8 | 923 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK154170.1, BC029153.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
