Assay Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081360.1 | NP_001074829.1 | 17 - 18 | 4682 | 64 | |
GenBank mRNA | AK081975.1 | - | 3 - 4 | 298 | 64 | |
AK122436.1 | - | 5 - 6 | 2022 | 64 | ||
BC138367.1 | - | 16 - 17 | 4258 | 8861387 | 64 | |
BC145258.1 | - | 15 - 16 | 4225 | 9053811 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
