Assay Details
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081011.2 | NP_001074480.2 | 15 - 16 | 2348 | 112 | |
GenBank mRNA | AK161932.1 | - | 4 - 5 | 252 | 112 | |
AY057900.1 | - | 6 - 7 | 337 | 112 | ||
BC030457.1 | - | 14 - 15 | 1655 | 112 | ||
BC151081.1 | - | 14 - 15 | 1966 | 9087994 | 112 | |
BC151082.1 | - | 14 - 15 | 1966 | 9087995 | 112 | |
BC158055.1 | - | 14 - 15 | 1964 | 9088163 | 112 | |
BC172152.1 | - | 14 - 15 | 1966 | 9054822 | 112 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK161932.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
