Assay Details
Gene Symbol: Traf3ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028718.2 | NP_082994.1 | 1 - 2 | 224 | 67 | |
GenBank mRNA | AY613437.1 | - | 1 - 2 | 227 | 67 | |
BC117868.2 | - | 1 - 2 | 198 | 40095476 | 67 | |
BY739359.1 | - | 1 - 2 | 224 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
