Assay Details
Gene Symbol: Cntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177129.5 | NP_796103.2 | 2 - 3 | 381 | 80 | |
GenBank mRNA | AK147573.1 | - | 2 - 3 | 307 | 80 | |
AK147602.1 | - | 2 - 3 | 382 | 80 | ||
AK147658.1 | - | 2 - 3 | 307 | 80 | ||
AK147673.1 | - | 2 - 3 | 332 | 80 | ||
BC066106.1 | - | 2 - 3 | 301 | 30362931 | 80 | |
CJ098637.1 | - | 2 - 3 | 382 | 80 | ||
CJ108593.1 | - | 2 - 3 | 382 | 80 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK147673.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
