Assay Details
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159965.1 | NP_001153437.1 | 3 - 4 | 383 | 91 | |
NM_001159966.1 | NP_001153438.1 | 3 - 4 | 281 | 91 | ||
NM_001159967.1 | NP_001153439.1 | 3 - 4 | 383 | 91 | ||
NM_023884.4 | NP_076373.3 | 3 - 4 | 323 | 91 | ||
NR_027640.1 | 3 - 4 | 383 | 91 | |||
GenBank mRNA | AF312924.1 | - | 2 - 3 | 231 | 91 | |
AK018622.1 | - | 3 - 4 | 325 | 91 | ||
AK029049.1 | - | 3 - 4 | 324 | 91 | ||
AK029993.1 | - | 3 - 4 | 356 | 91 | ||
AK033549.1 | - | 3 - 4 | 279 | 91 | ||
AK161080.1 | - | 3 - 4 | 350 | 91 | ||
AK165002.1 | - | 3 - 4 | 275 | 91 | ||
AK169368.1 | - | 3 - 4 | 301 | 91 | ||
BC043132.1 | - | 3 - 4 | 288 | 6811441 | 91 | |
BC052663.1 | - | 3 - 4 | 300 | 30051209 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
