Assay Details
Gene Symbol: 4933439K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040558.1 | 2 - 3 | 386 | 78 | ||
GenBank mRNA | AK017125.1 | - | 2 - 3 | 388 | 78 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY716803.1 | - | 386 | Chr. - - [-] on Build | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 37125 |
Gm37125 | -22.6kb | Anti-sense | Chr. 1 - 172512019 - 172518985 on NCBI Build GRCm38 | |
Downstream | V-set and immunoglobulin domain containing 8 | Vsig8 |
A030011M19, EG240916 |
12.9kb | Anti-sense | Chr. 1 - 172555938 - 172563717 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
