Assay Details
Gene Symbol: Usp9x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009481.2 | NP_033507.2 | 37 - 38 | 6827 | 84 | |
GenBank mRNA | AK034941.1 | - | 3 - 4 | 888 | 84 | |
DQ086491.1 | - | 37 - 38 | 6788 | 84 | ||
U67874.1 | - | 37 - 38 | 6788 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
