Assay Details
Gene Symbol: Gcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027375.2 | NP_081651.2 | 12 - 13 | 3472 | 98 | |
GenBank mRNA | AB093232.1 | - | 12 - 13 | 3475 | 98 | |
AK148283.1 | - | 7 - 8 | 687 | 98 | ||
BC023811.1 | - | 7 - 8 | 859 | 98 | ||
BC034077.1 | - | 1 - 2 | 110 | 98 | ||
BC139007.1 | - | 12 - 13 | 3468 | 8862029 | 98 | |
BC145495.1 | - | 12 - 13 | 3541 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
