Assay Details
Gene Symbol: Cog6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026225.3 | NP_080501.2 | 15 - 16 | 1603 | 75 | |
GenBank mRNA | AB093286.1 | - | 15 - 16 | 1575 | 75 | |
AK004100.1 | - | 1 - 2 | 84 | 75 | ||
AK016663.1 | - | 15 - 16 | 1615 | 75 | ||
AK031149.1 | - | 15 - 16 | 1597 | 75 | ||
AK045223.1 | - | 15 - 16 | 1597 | 75 | ||
AK153320.1 | - | 15 - 16 | 1592 | 75 | ||
AK218272.1 | - | 1 - 2 | 83 | 75 | ||
BC025427.1 | - | 15 - 16 | 1609 | 2649589 | 75 | |
BC071236.1 | - | 2 - 3 | 1303 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
