Assay Details
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172678.3 | NP_766266.3 | 4 - 5 | 569 | 85 | |
GenBank mRNA | AK031820.1 | - | 4 - 5 | 501 | 85 | |
AK075984.1 | - | 4 - 5 | 488 | 85 | ||
AK145423.1 | - | 4 - 5 | 477 | 85 | ||
AK189876.1 | - | 2 - 3 | 244 | 85 | ||
AK192710.1 | - | 3 - 4 | 232 | 86 | ||
AK195232.1 | - | 3 - 4 | 250 | 85 | ||
AK200479.1 | - | 3 - 4 | 231 | 85 | ||
AK205159.1 | - | 3 - 4 | 231 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
