Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 11 - 12 | 1354 | 83 | |
NM_001177626.1 | NP_001171097.1 | 11 - 12 | 1392 | 83 | ||
NM_007900.3 | NP_031926.2 | 12 - 13 | 1447 | 83 | ||
GenBank mRNA | AK040174.1 | - | 11 - 12 | 1327 | 83 | |
AK157718.1 | - | 11 - 12 | 1346 | 83 | ||
AK220452.1 | - | 11 - 12 | 1353 | 83 | ||
BC023881.1 | - | 11 - 12 | 1352 | 5354733 | 83 | |
BC025565.1 | - | 12 - 13 | 1421 | 5322744 | 83 | |
BC032155.1 | - | 11 - 12 | 1349 | 5067358 | 83 | |
BC045614.1 | - | 11 - 12 | 1327 | 5059799 | 83 | |
L11316.1 | - | 11 - 12 | 1292 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
