Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177626.1 | NP_001171097.1 | 1 - 2 | 180 | 81 | |
GenBank mRNA | AK040174.1 | - | 1 - 2 | 115 | 81 | |
AK157718.1 | - | 1 - 2 | 134 | 81 | ||
AK220452.1 | - | 1 - 2 | 141 | 81 | ||
BC023881.1 | - | 1 - 2 | 140 | 5354733 | 81 | |
BC045614.1 | - | 1 - 2 | 115 | 5059799 | 81 | |
BY070590.1 | - | 1 - 2 | 180 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK040174.1, AK157718.1, BC023881.1, BC045614.1, NM_001177626.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
