Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 2 - 3 | 296 | 70 | |
NM_001177626.1 | NP_001171097.1 | 2 - 3 | 334 | 70 | ||
NM_007900.3 | NP_031926.2 | 2 - 3 | 296 | 70 | ||
GenBank mRNA | AK040174.1 | - | 2 - 3 | 269 | 70 | |
AK157718.1 | - | 2 - 3 | 288 | 70 | ||
AK220452.1 | - | 2 - 3 | 295 | 70 | ||
BC023881.1 | - | 2 - 3 | 294 | 5354733 | 70 | |
BC025565.1 | - | 2 - 3 | 270 | 5322744 | 70 | |
BC045614.1 | - | 2 - 3 | 269 | 5059799 | 70 | |
BY070590.1 | - | 2 - 3 | 334 | 70 | ||
CF735294.1 | - | 1 - 2 | 168 | 70 | ||
CN702302.1 | - | 2 - 3 | 296 | 70 | ||
L11316.1 | - | 2 - 3 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L11316.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
