Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 4 - 5 | 560 | 93 | |
NM_001177626.1 | NP_001171097.1 | 4 - 5 | 598 | 93 | ||
NM_007900.3 | NP_031926.2 | 5 - 6 | 653 | 93 | ||
GenBank mRNA | AK040174.1 | - | 4 - 5 | 533 | 93 | |
AK157718.1 | - | 4 - 5 | 552 | 93 | ||
AK220452.1 | - | 4 - 5 | 559 | 93 | ||
BC023881.1 | - | 4 - 5 | 558 | 5354733 | 93 | |
BC025565.1 | - | 5 - 6 | 627 | 5322744 | 93 | |
BC045614.1 | - | 4 - 5 | 533 | 5059799 | 93 | |
CF735294.1 | - | 4 - 5 | 525 | 93 | ||
CN702302.1 | - | 4 - 5 | 560 | 93 | ||
L11316.1 | - | 4 - 5 | 498 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the L11316.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
