Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 14 - 15 | 1682 | 78 | |
NM_001177626.1 | NP_001171097.1 | 14 - 15 | 1720 | 78 | ||
NM_007900.3 | NP_031926.2 | 15 - 16 | 1775 | 78 | ||
GenBank mRNA | AK040174.1 | - | 14 - 15 | 1655 | 78 | |
AK157718.1 | - | 14 - 15 | 1674 | 78 | ||
AK220452.1 | - | 14 - 15 | 1681 | 78 | ||
BC023881.1 | - | 14 - 15 | 1680 | 5354733 | 78 | |
BC025565.1 | - | 15 - 16 | 1749 | 5322744 | 78 | |
BC032155.1 | - | 14 - 15 | 1677 | 5067358 | 78 | |
BC045614.1 | - | 14 - 15 | 1655 | 5059799 | 78 | |
L11316.1 | - | 14 - 15 | 1620 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
