Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 16 - 17 | 1881 | 100 | |
NM_001177626.1 | NP_001171097.1 | 16 - 17 | 1919 | 100 | ||
NM_007900.3 | NP_031926.2 | 17 - 18 | 1974 | 100 | ||
GenBank mRNA | AK040174.1 | - | 16 - 17 | 1854 | 100 | |
AK157718.1 | - | 16 - 17 | 1873 | 100 | ||
AK220452.1 | - | 16 - 17 | 1880 | 100 | ||
BC023881.1 | - | 16 - 17 | 1879 | 5354733 | 100 | |
BC025565.1 | - | 17 - 18 | 1948 | 5322744 | 100 | |
BC032155.1 | - | 16 - 17 | 1876 | 5067358 | 100 | |
BC045614.1 | - | 16 - 17 | 1854 | 5059799 | 100 | |
L11316.1 | - | 16 - 17 | 1819 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
