Assay Details
Gene Symbol: Ect2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177625.1 | NP_001171096.1 | 1 - 2 | 139 | 67 | |
NM_007900.3 | NP_031926.2 | 1 - 2 | 139 | 67 | ||
GenBank mRNA | BC025565.1 | - | 1 - 2 | 113 | 5322744 | 67 |
CN702302.1 | - | 1 - 2 | 139 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC025565.1, NM_001177625.1, NM_007900.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
