Assay Details
Gene Symbol: Slc33a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272035.1 | NP_001258964.1 | 1 - 2 | 1315 | 156 | |
NM_015728.5 | NP_056543.2 | 1 - 2 | 1315 | 156 | ||
GenBank mRNA | AK144703.1 | - | 1 - 2 | 1181 | 157 | |
BC005616.1 | - | 1 - 2 | 892 | 3154886 | 156 | |
BC054395.1 | - | 1 - 2 | 1315 | 6492236 | 156 | |
BC063104.1 | - | 1248 | 5042025 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
