Assay Details
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033565.2 | NP_291043.1 | 3 - 4 | 1273 | 136 | |
GenBank mRNA | AF190449.1 | - | 3 - 4 | 1215 | 136 | |
AK044058.1 | - | 3 - 4 | 1157 | 136 | ||
AK054401.1 | - | 3 - 4 | 1121 | 136 | ||
BC138999.1 | - | 3 - 4 | 946 | 8862021 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
