Assay Details
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033565.2 | NP_291043.1 | 6 - 7 | 1430 | 81 | |
GenBank mRNA | AF190449.1 | - | 6 - 7 | 1372 | 81 | |
AK044058.1 | - | 6 - 7 | 1314 | 81 | ||
AK054401.1 | - | 6 - 7 | 1278 | 81 | ||
BC138999.1 | - | 6 - 7 | 1103 | 8862021 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
