Assay Details
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013919.4 | NP_038947.2 | 12 - 13 | 1834 | 60 | |
GenBank mRNA | AF177759.1 | - | 11 - 12 | 1633 | 60 | |
AK011148.1 | - | 12 - 13 | 1844 | 60 | ||
AK013252.1 | - | 11 - 12 | 1750 | 60 | ||
AK207403.1 | - | 1 - 2 | 111 | 60 | ||
BC021903.1 | - | 12 - 13 | 1824 | 3985792 | 60 | |
KF646669.1 | - | 11 - 12 | 1352 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
ubiquitin-dependent protein catabolic process
histone deubiquitination
neuron projection development
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription, DNA-templated
regulation of proteasomal protein catabolic process
transcription coactivator activity
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
peptidase activity
cysteine-type peptidase activity
hydrolase activity
NEDD8-specific protease activity
metal ion binding
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
ubiquitin-dependent protein catabolic process
histone deubiquitination
neuron projection development
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of transcription, DNA-templated
regulation of proteasomal protein catabolic process
transcription coactivator activity
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
peptidase activity
cysteine-type peptidase activity
hydrolase activity
NEDD8-specific protease activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827148 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827153 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827150 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827149 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827152 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827151 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067181 | AK013252.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067181 | BC021903.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067181 | NM_013919.4 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067181 | AK011148.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2067181 | BP770216.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8689 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150766 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450058 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131938 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067181 | AK013252.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067181 | BC021903.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067181 | NM_013919.4 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067181 | AK011148.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2067181 | BP770216.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827119 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827111 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827109 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827154 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827114 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827116 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827148 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827133 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827120 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827142 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827145 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827151 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00250713 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131938 | - | - |
Whole Mouse Genome G4122A | A_51_P346747 | AK013252.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P346747 | AK207403.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P346747 | BC021903.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P346747 | NM_013919.4 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P346747 | AF177759.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116847 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131938 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00289290 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00289255 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00481671 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00289233 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827149 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827153 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827148 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827152 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827151 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827114 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00853176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827119 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00481648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289255 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827108 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00168121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289276 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827150 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289269 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289242 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01009891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827129 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827153 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827127 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827121 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00289240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20827118 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00481636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10360113 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360117 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360118 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360108 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360115 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360112 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360110 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360107 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360114 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360119 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360111 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360116 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360106 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10360109 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454034_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10360105 | - | - |
Mouse Genome 430 2.0 Array | 1454034_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1454034_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30941_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 30941_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0100028971.mm.1 | TC0100003537.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100028971.mm.1 | NM_013919.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100028971.mm.1 | NM_013919.4 | Refseq |
Mouse Expression Set 430 | 1454034_a_at | - | - |
Mouse Expression Set 430 | 1454034_PM_a_at | - | - |
Murine Genome U74v2 Set | 95027_at | - | - |
Murine Genome U74v2 Set | 95027_PM_at | - | - |
Murine Genome U74v2 Set | 137510_PM_at | - | - |
Murine Genome U74v2 Set | 167175_PM_at | - | - |
Murine Genome U74v2 Set | 167175_at | - | - |
Murine Genome U74v2 Set | 137510_at | - | - |
Mouse Genome 430A 2.0 Array | 1454034_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1454034_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1454034_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1454034_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 656183 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3060446 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3060446 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101850301 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003060446 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2495939 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1254123 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2495939 | - | - |
mouse-6 v1.1 (Illumina) | scl15944.6.1_77-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3060446 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101850301 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl15944.6.1_77-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3060446 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101850301 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2495939 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254123 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2495939 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl15944.6.1_77-S | - | - |
