Assay Details
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081756.1 | NP_001075225.1 | 14 - 15 | 5376 | 61 | |
NM_172484.3 | NP_766072.2 | 12 - 13 | 5237 | 61 | ||
NM_176957.3 | NP_795931.2 | 11 - 12 | 1266 | 61 | ||
GenBank mRNA | AK053243.1 | - | 9 - 10 | 949 | 61 | |
AY957601.1 | - | 9 - 10 | 4604 | 61 | ||
AY957602.1 | - | 10 - 11 | 4796 | 61 | ||
BC066095.1 | - | 12 - 13 | 5237 | 6856184 | 61 | |
BC079551.1 | - | 12 - 13 | 5237 | 6856184 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
