Assay Details
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081756.1 | NP_001075225.1 | 1 - 2 | 322 | 91 | |
NM_176957.3 | NP_795931.2 | 1 - 2 | 322 | 91 | ||
GenBank mRNA | AK041950.1 | - | 1 - 2 | 172 | 91 | |
CN700744.1 | - | 1 - 2 | 332 | 91 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK041950.1, NM_001081756.1, NM_176957.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
