Assay Details
Gene Symbol: Iqca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029122.2 | NP_083398.2 | 14 - 15 | 1893 | 76 | |
GenBank mRNA | AK139726.1 | - | 7 - 8 | 748 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
