Assay Details
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173398.3 | NP_775574.1 | 1 | 163 | ||
GenBank mRNA | AK041381.1 | - | 1 | 163 | ||
AK041582.1 | - | 1 | 163 | |||
AK079674.1 | - | 1 | 163 | |||
AK083323.1 | - | 1 | 163 | |||
AK089252.1 | - | 1 | 163 | |||
AK154725.1 | - | 1 | 163 | |||
BC024054.1 | - | 2 | 5068166 | 128 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK041381.1,AK041582.1,AK089252.1,AK154725.1,BC024054.1,NM_173398.3 transcripts.
This assay is in the 5'UTR region of the AK041381.1,AK041582.1,AK089252.1,AK154725.1,BC024054.1,NM_173398.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
