Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100010929.mm.1 |
TC0100001316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010929.mm.1 |
NM_001038592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010929.mm.1 |
NM_001038592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010929.mm.1 |
NM_023505.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010929.mm.1 |
NM_023505.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010931.mm.1 |
TC0100001316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010931.mm.1 |
NM_001038592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010931.mm.1 |
NM_001038592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
TC0100001316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038594.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_001038594.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_023505.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010934.mm.1 |
NM_023505.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010926.mm.1 |
TC0100001316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010926.mm.1 |
NM_001038592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010926.mm.1 |
NM_001038592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010930.mm.1 |
TC0100001316.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010930.mm.1 |
NM_001038592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010930.mm.1 |
NM_001038592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010930.mm.1 |
NM_023505.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010930.mm.1 |
NM_023505.2 |
Refseq |