Assay Details
Gene Symbol: Lgr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC026896.1 | - | 2 - 3 | 126 | 84 | |
BC141210.1 | - | 17 - 18 | 1736 | 9055822 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
