Assay Details
Gene Symbol: Gm12532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081291.1 | - | 2 - 3 | 575 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | interleukin 2 | Il2 | Il-2 | -58.0kb | Anti-sense | Chr. 3 - 37120713 - 37125954 on NCBI Build GRCm38 |
Downstream | interleukin 21 | Il21 | IL-21 | 36.3kb | Anti-sense | Chr. 3 - 37222759 - 37232636 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
