Assay Details
Gene Symbol: Bcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009741.4 | NP_033871.2 | 1 - 2 | 1989 | 85 | |
GenBank mRNA | AK049473.1 | - | 1 - 2 | 1983 | 85 | |
BC095964.1 | - | 1 - 2 | 1645 | 6306635 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
