Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 10 - 11 | 1901 | 136 | ||
GenBank mRNA | AK006249.1 | - | 8 - 9 | 1167 | 136 | |
AY461596.1 | - | 10 - 11 | 1919 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() |
