Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 2 - 3 | 702 | 169 | ||
GenBank mRNA | AY461596.1 | - | 2 - 3 | 720 | 169 |

More Information
Set Membership: |
![]() ![]() ![]() |
