Assay Details
Gene Symbol: Slco6b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120500.1 | 3 - 4 | 895 | 109 | ||
GenBank mRNA | AK006249.1 | - | 1 - 2 | 161 | 109 | |
AK016463.1 | - | 3 - 4 | 907 | 109 | ||
AY461596.1 | - | 3 - 4 | 913 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
